Learn More
Med Vet International Oasis Monofyl Suture, Size 4-0, with NFS-2 Needle, 12/Box, Veterinary Use Only
Catalog No. 501180605
Encompass_Preferred
The Oasis veterinary use Monofyl suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ Monocryl®. The Monofyl suture has an absorption rate between 91 and 119 days. Size: 4-0. Needle Type: NFS-2, 19mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box
Catalog No. 50-118-0605 Supplier Med Vet International Supplier No. MVY922V
The Oasis veterinary use Monofyl suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ Monocryl®. The Monofyl suture has an absorption rate between 91 and 119 days. Size: 4-0. Needle Type: NFS-2, 19mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box