Learn More
Fine Science Tools SUTURE NEEDLES SIZE 0 12PK
SDP
Catalog No. NC9575549
Encompass
Product: Kalt Suture Needles - Size 0/ Size: 0/ Needle Diameter: 0.35mm/ Needle Circle: 3/8/ Needle Arch: 22.4mm/ Needle Chord: 17.5mm/ Needle Alloy: Stainless Steel/ Sterile: No/ Unit: Dozen
Catalog No. NC9575549 Supplier Fine Science Tools Supplier No. 1205010
Product: Kalt Suture Needles - Size 0/ Size: 0/ Needle Diameter: 0.35mm/ Needle Circle: 3/8/ Needle Arch: 22.4mm/ Needle Chord: 17.5mm/ Needle Alloy: Stainless Steel/ Sterile: No/ Unit: Dozen