Learn More
Med Vet International Oasis Nylon Suture, Size 4-0, with NFS-2 Needle, 12/Box, Veterinary Use Only
Catalog No. 501180629
Encompass_Preferred
The veterinary use Oasis Nylon suture with needle is a monofilament non-absorbable suture that is comparable to Ethicon™ Ethilon®. This Nylon suture has extremely high tensile strength that exceeds USP requirements. It provides excellent knot security, while the smooth surface allows for easy passage through the tissue. The thread color of this suture is black and has a composition of polyamide. Size: 4-0. Needle Type: NFS-2, 19mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box
Catalog No. 50-118-0629 Supplier Med Vet International Supplier No. MV662V
The veterinary use Oasis Nylon suture with needle is a monofilament non-absorbable suture that is comparable to Ethicon™ Ethilon®. This Nylon suture has extremely high tensile strength that exceeds USP requirements. It provides excellent knot security, while the smooth surface allows for easy passage through the tissue. The thread color of this suture is black and has a composition of polyamide. Size: 4-0. Needle Type: NFS-2, 19mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box