Learn More
Med Vet International Oasis Nylon Suture Cassette, Size 3, Length of 50M, Each
Catalog No. 502643564
Encompass_Preferred
The veterinary use Oasis Nylon suturecassette is a monofilament non-absorbable suture. This Nylon suture has extremely high tensile strength that exceeds USP requirements. It provides excellent knot security, while the smooth surface allows for easy passage through the tissue. The thread color of this suture is blue and has a composition of polyamide. Size: 3. Length: 50M.
Catalog No. 50-264-3564 Supplier Med Vet International Supplier No. MF03
The veterinary use Oasis Nylon suturecassette is a monofilament non-absorbable suture. This Nylon suture has extremely high tensile strength that exceeds USP requirements. It provides excellent knot security, while the smooth surface allows for easy passage through the tissue. The thread color of this suture is blue and has a composition of polyamide. Size: 3. Length: 50M.