Learn More
Med Vet International Oasis PDO Suture, Size 4-0, with NSH Needle, 12/Box, Veterinary Use Only
Catalog No. 501180673
Encompass_Preferred
The veterinary use Oasis PDO suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ PDS®. The PDO smooth monofilament suture has a gradual absorption rate between 180 and 220 days. It retains 50% of its tensile strength for up to 6 weeks. This suture provides excellent pliability and holding strength. Size: 4-0. Needle Type: NSH, 26mm 1/2 Circle Taper Point. Length: 30in. Quantity: 12/box
Catalog No. 50-118-0673 Supplier Med Vet International Supplier No. MVZ315V
The veterinary use Oasis PDO suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ PDS®. The PDO smooth monofilament suture has a gradual absorption rate between 180 and 220 days. It retains 50% of its tensile strength for up to 6 weeks. This suture provides excellent pliability and holding strength. Size: 4-0. Needle Type: NSH, 26mm 1/2 Circle Taper Point. Length: 30in. Quantity: 12/box