Learn More
Med Vet International Oasis PDO Suture Cassette, Size 2, Length of 15M, Each
Catalog No. 502643463
Encompass_Preferred
The veterinary use Oasis PDO suture cassette is a monofilament synthetic absorbable suture. The PDO smooth monofilament suture has a gradual absorption rate between 180 and 220 days. It retains 50% of its tensile strength for up to 6 weeks. This suture provides excellent pliability and holding strength. Size: 2. Length: 15M.
Catalog No. 50-264-3463 Supplier Med Vet International Supplier No. PD02
The veterinary use Oasis PDO suture cassette is a monofilament synthetic absorbable suture. The PDO smooth monofilament suture has a gradual absorption rate between 180 and 220 days. It retains 50% of its tensile strength for up to 6 weeks. This suture provides excellent pliability and holding strength. Size: 2. Length: 15M.