Learn More
Med Vet International Ethicon PDS II Suture, Size 2-0, FS-1, 27in, 36/Box
Catalog No. 502522110
Encompass_Preferred
The Ethicon Z443H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 2-0, they are dyed undyed to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a FS-1 reverse cutting needle and has 36 sutures in a box.
Catalog No. 50-252-2110 Supplier Med Vet International Supplier No. Z443H
The Ethicon Z443H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 2-0, they are dyed undyed to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a FS-1 reverse cutting needle and has 36 sutures in a box.