Learn More
Med Vet International Ethicon PDS II Suture, Size 3-0, FS-2, 27in, 36/Box
Catalog No. NC1985435
Encompass_Preferred
The Ethicon Z423H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed undyed to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a FS-2 reverse cutting needle and has 36 sutures in a box.
This item is not returnable. View return policy
Catalog No. NC1985435 Supplier Med Vet International Supplier No. Z423H
The Ethicon Z423H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed undyed to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a FS-2 reverse cutting needle and has 36 sutures in a box.