Learn More
Med Vet International Ethicon PDS II Suture, Size 4-0, PS-2, 18in, Clear, 12/Box
Catalog No. 501180705
Encompass_Preferred
The Ethicon Z496G suture is a high-quality absorbable Polydioxanone surgical suture. It measures 18in. in length, has a size of 4-0, they are undyed (clear), and typically have an absorption rate of 182-238 days. This suture comes with a PS-2 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-118-0705 Supplier Med Vet International Supplier No. Z496G
The Ethicon Z496G suture is a high-quality absorbable Polydioxanone surgical suture. It measures 18in. in length, has a size of 4-0, they are undyed (clear), and typically have an absorption rate of 182-238 days. This suture comes with a PS-2 Precision Point reverse cutting needle and has 12 sutures in a box.