Learn More
Med Vet International Ethicon PDS II Suture, Size 4-0, RB-1, 27in, 36/Box
Catalog No. 501180706
Encompass_Preferred
The Ethicon Z304H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 4-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a RB-1 taper point needle and has 36 sutures in a box.
Catalog No. 50-118-0706 Supplier Med Vet International Supplier No. Z304H
The Ethicon Z304H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 4-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a RB-1 taper point needle and has 36 sutures in a box.