Learn More
Med Vet International Ethicon PDS Suture 4-0, P-3 needle, 18in, Clear, Qty/bx: 12
Catalog No. 502522106
Encompass_Preferred
The Ethicon Z494G suture is a high-quality absorbable Polydioxanone surgical suture. It measures 18in. in length, has a size of 4-0, they are undyed (clear), and typically have an absorption rate of 182-238 days. This suture comes with a P3 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2106 Supplier Med Vet International Supplier No. Z494G
The Ethicon Z494G suture is a high-quality absorbable Polydioxanone surgical suture. It measures 18in. in length, has a size of 4-0, they are undyed (clear), and typically have an absorption rate of 182-238 days. This suture comes with a P3 Precision Point reverse cutting needle and has 12 sutures in a box.