Learn More
Med Vet International Ethicon Pds Ii Polydioxanone Suture, Size 0, CTX, 36in, Violet, 24/Box
Catalog No. 502522111
Encompass_Preferred
The Ethicon Z370T suture is a high-quality absorbable Pds Ii Polydioxanone suture. It measures 36in in length, has a size of 0, and are dyed violet to provide greater visibility while suturing. This suture comes with a CTX 48mm 1/2c Taper Needle and has 24 sutures in a box.
Catalog No. 50-252-2111 Supplier Med Vet International Supplier No. Z370T
The Ethicon Z370T suture is a high-quality absorbable Pds Ii Polydioxanone suture. It measures 36in in length, has a size of 0, and are dyed violet to provide greater visibility while suturing. This suture comes with a CTX 48mm 1/2c Taper Needle and has 24 sutures in a box.