Learn More
Med Vet International Ethicon PDS Plus Antibacterial Suture, Size 0, CT-2, 27in, 36/Box
Supplier:  Med Vet International PDP334H
Encompass_Preferred
The Ethicon PDS Plus PDP334H suture is a high-quality absorbable Polydioxanone surgical suture. It measures 27in. in length, has a size of 0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 182-238 days. This suture comes with a CT-2 taper point needle and has 36 sutures in a box.
Catalog No. 50-252-2200