Learn More
Med Vet International Ethicon Permahand Silk Suture, Size 3-0, SH, 30in, Black, 36/Box
Catalog No. 502522326
Encompass_Preferred
The Ethicon K832H suture is a high-quality non-absorbable Permahand Silk suture. It measures 30in in length, has a size of 3-0, and are dyed black to provide greater visibility while suturing. This suture comes with a SH 26mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-252-2326 Supplier Med Vet International Supplier No. K832H
The Ethicon K832H suture is a high-quality non-absorbable Permahand Silk suture. It measures 30in in length, has a size of 3-0, and are dyed black to provide greater visibility while suturing. This suture comes with a SH 26mm 1/2c Taper Needle and has 36 sutures in a box.