Learn More
Med Vet International Oasis PGA Suture, Size 2-0, with NCP-1 Needle, 12/Box, Veterinary Use Only
Catalog No. 502522252
Encompass_Preferred
The veterinary use Oasis PGA suture with needle is a braided synthetic absorbable suture that is comparable to Ethicon™ Vicryl®. The PGA braided suture has a rapid absorption rate between 60 and 90 days. It retains 60 to 70% of its tensile strength at 14 days. This suture provides excellent pliability and holding strength. Size: 2-0. Needle Type: NCP-1, 36mm 1/2 Circle Reverse Cutting Point. Length: 30in. Quantity: 12/box
Catalog No. 50-252-2252 Supplier Med Vet International Supplier No. MVJ466V
The veterinary use Oasis PGA suture with needle is a braided synthetic absorbable suture that is comparable to Ethicon™ Vicryl®. The PGA braided suture has a rapid absorption rate between 60 and 90 days. It retains 60 to 70% of its tensile strength at 14 days. This suture provides excellent pliability and holding strength. Size: 2-0. Needle Type: NCP-1, 36mm 1/2 Circle Reverse Cutting Point. Length: 30in. Quantity: 12/box