Learn More
Med Vet International Oasis PGA Suture, Size 3-0, with NFS-1 Needle, 12/Box
Catalog No. 502522253
Encompass_Preferred
The Oasis PGA suture with needle is a braided synthetic absorbable suture that is comparable to Ethicon™ Vicryl®. The PGA braided suture has a rapid absorption rate between 60 and 90 days. It retains 60 to 70% of its tensile strength at 14 days. This suture provides excellent pliability and holding strength. Size: 3-0. Needle Type: NFS-1, 24mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box
Catalog No. 50-252-2253 Supplier Med Vet International Supplier No. MVJ452
The Oasis PGA suture with needle is a braided synthetic absorbable suture that is comparable to Ethicon™ Vicryl®. The PGA braided suture has a rapid absorption rate between 60 and 90 days. It retains 60 to 70% of its tensile strength at 14 days. This suture provides excellent pliability and holding strength. Size: 3-0. Needle Type: NFS-1, 24mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box