Learn More
Med Vet International Oasis PGA Suture Cassette, Size 3-0, Length of 15M, Each
Catalog No. 502643458
Encompass_Preferred
The veterinary use Oasis PGA suture cassette is a braided synthetic absorbable suture. The PGA braided suture has a rapid absorption rate between 60 and 90 days. It retains 60 to 70% of its tensile strength at 14 days. This suture provides excellent pliability and holding strength. Size: 3-0. Length: 15M.
Catalog No. 50-264-3458 Supplier Med Vet International Supplier No. PG30
The veterinary use Oasis PGA suture cassette is a braided synthetic absorbable suture. The PGA braided suture has a rapid absorption rate between 60 and 90 days. It retains 60 to 70% of its tensile strength at 14 days. This suture provides excellent pliability and holding strength. Size: 3-0. Length: 15M.