Learn More
Med Vet International Ethicon Plain Gut Suture 5-0, PC-1 needle 18in Fast Absorbing 12/box
Catalog No. 502522576
Encompass_Preferred
The Ethicon1915G suture is a high-quality fast absorbing Plain Gut surgical suture. It measures 18 in. length, size of 5-0, Yellowish Tan to provide visibility while suturing, and typically have an absorption rate of 21-42 days. This suture comes with a PC-1 Precision Point conventional cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2576 Supplier Med Vet International Supplier No. 1915G
The Ethicon1915G suture is a high-quality fast absorbing Plain Gut surgical suture. It measures 18 in. length, size of 5-0, Yellowish Tan to provide visibility while suturing, and typically have an absorption rate of 21-42 days. This suture comes with a PC-1 Precision Point conventional cutting needle and has 12 sutures in a box.