Learn More
Med Vet International Ethicon Plain Gut 6-0, G-1 needle, 18in, 12/bpx
Catalog No. 502522517
Encompass_Preferred
The Ethicon 774G suture is a high-quality absorbable Plain Gut surgical suture. It measures 18in. in length, has a size of 6-0, yellowish tan to provide greater visibility while suturing, This suture comes with a G-1 reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2517 Supplier Med Vet International Supplier No. 774G
The Ethicon 774G suture is a high-quality absorbable Plain Gut surgical suture. It measures 18in. in length, has a size of 6-0, yellowish tan to provide greater visibility while suturing, This suture comes with a G-1 reverse cutting needle and has 12 sutures in a box.