Description
- Realistic multi-layer silicone suture pad with pre-cut wound patterns for practicing various techniques.
- Premium stainless-steel instruments including needle holder, Adson forceps, scissors, and scalpel handle.
- Includes mixed surgical sutures for repeated practice sessions.
- Reusable, durable materials designed to withstand multiple training sessions without tearing.
- Portable zippered case keeps tools organized and ready for lab, class, or home practice.
- Ideal for students and professionals learning suturing, knot-tying, and incision handling.
- 1 x Silicone Suture Practice Pad (multi-wound)
- 1 x Iris Scissors
- 1 x Stitch Scissors
- 1 x Needle Holder
- 1 x Adson Forceps
- 1 x Scalpel Handle
- Mixed Sutures (variety pack)
- Spare scalpel blades
- Zippered instrument case
Specifications
Specifications
| Material | Stainless Steel |
| Autoclavable | Autoclavable |
| For Use With (Application) | Learning, Training, Education |
| Includes | Suture Variety Pack |
| Sterility | Non-sterile |
| Product Type | Suture Laceration Set |