Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 0, MO-6, 30in, Blue, 36/Box
Catalog No. 502522504
Encompass_Preferred
The Ethicon 8418H suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 30in in length, has a size of 0, and are dyed blue to provide greater visibility while suturing. This suture comes with a MO-6 26mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-252-2504 Supplier Med Vet International Supplier No. 8418H
The Ethicon 8418H suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 30in in length, has a size of 0, and are dyed blue to provide greater visibility while suturing. This suture comes with a MO-6 26mm 1/2c Taper Needle and has 36 sutures in a box.