Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 1, CT-1, 30in, 36/Box
Catalog No. 502522503
Encompass_Preferred
The Ethicon 8425H suture is a high-quality non-absorbable Polypropylene surgical suture. It measures 30in. in length, has a size of 1, and are dyed black to provide greater visibility while suturing. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.
Catalog No. 50-252-2503 Supplier Med Vet International Supplier No. 8425H
The Ethicon 8425H suture is a high-quality non-absorbable Polypropylene surgical suture. It measures 30in. in length, has a size of 1, and are dyed black to provide greater visibility while suturing. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.