Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 2-0, CT-2, 30in, Blue, 36/Box
Catalog No. 502522505
Encompass_Preferred
The Ethicon 8411H suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 30in in length, has a size of 2-0, and are dyed blue to provide greater visibility while suturing. This suture comes with a CT-2 26mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-252-2505 Supplier Med Vet International Supplier No. 8411H
The Ethicon 8411H suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 30in in length, has a size of 2-0, and are dyed blue to provide greater visibility while suturing. This suture comes with a CT-2 26mm 1/2c Taper Needle and has 36 sutures in a box.