Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 2-0, Double Armed RB-1 RB-1, 36in, Blue, 36/Box
Catalog No. 502522500
Encompass_Preferred
The Ethicon 8559H suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 36in in length, has a size of 2-0, and are dyed blue to provide greater visibility while suturing. This suture comes with a Double Armed RB-1 RB-1 17mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-252-2500 Supplier Med Vet International Supplier No. 8559H
The Ethicon 8559H suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 36in in length, has a size of 2-0, and are dyed blue to provide greater visibility while suturing. This suture comes with a Double Armed RB-1 RB-1 17mm 1/2c Taper Needle and has 36 sutures in a box.