Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 3-0, FS-2, 18in, 12/Box, 8665G
Catalog No. 502522497
Encompass_Preferred
The Ethicon 8665G suture is a high-quality non-absorbable Polypropylene surgical suture. It measures 18in. in length, has a size of 3-0, and are dyed black to provide greater visibility while suturing. This suture comes with a FS-2 reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-252-2497 Supplier Med Vet International Supplier No. 8665G
The Ethicon 8665G suture is a high-quality non-absorbable Polypropylene surgical suture. It measures 18in. in length, has a size of 3-0, and are dyed black to provide greater visibility while suturing. This suture comes with a FS-2 reverse cutting needle and has 12 sutures in a box.