Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 6-0, P-3, 18in, 12/Box
Catalog No. 501180794
Encompass_Preferred
The Ethicon 8695G suture is a high-quality non-absorbable Polypropylene surgical suture. It measures 18in. in length, has a size of 6-0, and are dyed black to provide greater visibility while suturing. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-118-0794 Supplier Med Vet International Supplier No. 8695G
The Ethicon 8695G suture is a high-quality non-absorbable Polypropylene surgical suture. It measures 18in. in length, has a size of 6-0, and are dyed black to provide greater visibility while suturing. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.