Learn More
Sklar Instruments Econo Littauer Suture Scissors, Sklar, 3-1/2 in., Blunt/Blunt, Scissors, Curved, Smooth, Stainless Steel. PK/12.
SDP
Catalog No. 501437613 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Littauer Suture Scissors, Sklar, 3-1/2 in., Blunt/Blunt, Scissors, Curved, Smooth, Stainless Steel. Econo Littauer Suture Scissors are a disposable, floor-grade instrument used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Littauer suture scissors typically have larger, less delicate tips than Spencer Stitch Scissors, and are often used for removing larger sutures. They should not be used for cutting tissue. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is curved with smooth, blunt/blunt tips and a length of 3-1/2 inches. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7613 Supplier Sklar Instruments Supplier No. 21346
Econo Littauer Suture Scissors, Sklar, 3-1/2 in., Blunt/Blunt, Scissors, Curved, Smooth, Stainless Steel. Econo Littauer Suture Scissors are a disposable, floor-grade instrument used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Littauer suture scissors typically have larger, less delicate tips than Spencer Stitch Scissors, and are often used for removing larger sutures. They should not be used for cutting tissue. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is curved with smooth, blunt/blunt tips and a length of 3-1/2 inches. They are sold in boxes of 12.