Learn More
Med Vet International Suture Screw/Anchor, 12mm L, 2.0mm eyelet dia, 2.5mm pilot hole, J0836F
Catalog No. 5000582005
Encompass_Preferred
Suture anchors have many uses, particularly reattachment of a ligament to bone. A screw is placed into bone and the head of the screw has an eyelet for suture placement. The screws are self-tapping after placement of a smaller pilot hole. A special driver is used to insert the screw. Suggested uses include:Coxo femoral luxationsShearing injuriesCollateral ligament repairsAnchor for CCL lateral sutures
Catalog No. 50-005-82005 Supplier Med Vet International Supplier No. JOR836F
Suture anchors have many uses, particularly reattachment of a ligament to bone. A screw is placed into bone and the head of the screw has an eyelet for suture placement. The screws are self-tapping after placement of a smaller pilot hole. A special driver is used to insert the screw. Suggested uses include:Coxo femoral luxationsShearing injuriesCollateral ligament repairsAnchor for CCL lateral sutures