Learn More
Med Vet International Suture Screw/Anchor, 8mm L. 1.5mm eyelet dia. 2.0mm pilot hole, J0836E
Catalog No. 5000581992
Encompass_Preferred
Suture anchors have many uses, particularly reattachment of a ligament to bone. A screw is placed into bone and the head of the screw has an eyelet for suture placement. The screws are self-tapping after placement of a smaller pilot hole. A special driver is used to insert the screw. Suggested uses include:Coxo femoral luxationsShearing injuriesCollateral ligament repairsAnchor for CCL lateral sutures
Catalog No. 50-005-81992 Supplier Med Vet International Supplier No. JOR836E
Suture anchors have many uses, particularly reattachment of a ligament to bone. A screw is placed into bone and the head of the screw has an eyelet for suture placement. The screws are self-tapping after placement of a smaller pilot hole. A special driver is used to insert the screw. Suggested uses include:Coxo femoral luxationsShearing injuriesCollateral ligament repairsAnchor for CCL lateral sutures