Learn More
McKesson General Medical SUTURE, SILK 4-0 C-6 (12/BX)
Catalog No. NC1862947
Encompass
Black Suture Braided Silk Size 4 - 0 18 Inch Suture 1-Needle 19 mm Length 3/8 Circle Reverse Cutting Needle
This item is not returnable. View return policy
Catalog No. NC1862947 Supplier McKesson General Medical Supplier No. 301688
Black Suture Braided Silk Size 4 - 0 18 Inch Suture 1-Needle 19 mm Length 3/8 Circle Reverse Cutting Needle