Learn More
Med Vet International Silk Suture Cassette, 1-0, 100m Veterinary Use Only
Catalog No. 502643440
Encompass_Preferred
Non-absorbable silk braided suture Extremely strong tensile strength High tensile strength suture that exceeds USP requirements Excellent knot security Designed for secure knotting Smooth surface allows for easy passage through tissue Composition: Silk Absorption Type: Non-Absorbable Structure: Braided Thread Color: Black
Catalog No. 50-264-3440 Supplier Med Vet International Supplier No. SLK1
Non-absorbable silk braided suture Extremely strong tensile strength High tensile strength suture that exceeds USP requirements Excellent knot security Designed for secure knotting Smooth surface allows for easy passage through tissue Composition: Silk Absorption Type: Non-Absorbable Structure: Braided Thread Color: Black