Learn More
Med Vet International Oasis Silk Suture, Size 6-0, with NC-3 Needle, 12/Box
Catalog No. 501180809
Encompass_Preferred
The Oasis Silk suture with needle is a braided non-absorbable suture. This sterile surgical suture is composed of an organic protein called fibroin. This protein is derived from the domesticated species Bombyx mori (B. mori) of the family Bombycidae. This line of sutures has FDA approved black dye for enhanced visiblity.Size: 6-0. Needle Type: NC-3. Length: 30in. Quantity: 12/box
Catalog No. 50-118-0809 Supplier Med Vet International Supplier No. MV711
The Oasis Silk suture with needle is a braided non-absorbable suture. This sterile surgical suture is composed of an organic protein called fibroin. This protein is derived from the domesticated species Bombyx mori (B. mori) of the family Bombycidae. This line of sutures has FDA approved black dye for enhanced visiblity.Size: 6-0. Needle Type: NC-3. Length: 30in. Quantity: 12/box