Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 0, Ligapak Reel, 54in, Violet, 12/Box
Catalog No. 502830753
Encompass_Preferred
The Ethicon VCP207G suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 54in in length, has a size of 3-0, and are violet. This suture comes with a Ligapak Reel needle and has 12 sutures in a box. Primary code substitute for discontinued code J207G
Catalog No. 50-283-0753 Supplier Med Vet International Supplier No. VCP207G
The Ethicon VCP207G suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 54in in length, has a size of 3-0, and are violet. This suture comes with a Ligapak Reel needle and has 12 sutures in a box. Primary code substitute for discontinued code J207G