Learn More
Med Vet International Ethicon Vicryl Suture, Size 0, CP-1, 27in, 36/Box
Catalog No. NC1985444
Encompass_Preferred
The Ethicon J467H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CP-1 reverse cutting needle and has 36 sutures in a box.
This item is not returnable. View return policy
Catalog No. NC1985444 Supplier Med Vet International Supplier No. J467H
The Ethicon J467H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CP-1 reverse cutting needle and has 36 sutures in a box.