Learn More
Med Vet International Ethicon Vicryl Suture, Size 0, CT-1, 36in, 36/Box
Catalog No. 502522361
Encompass_Preferred
The Ethicon J346H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 36in. in length, has a size of 0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.
Catalog No. 50-252-2361 Supplier Med Vet International Supplier No. J346H
The Ethicon J346H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 36in. in length, has a size of 0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.