Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 2-0, Cp-2, 27in, Undyed, 36/Box
Catalog No. 502830030
Encompass_Preferred
The Ethicon VCP869H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 2-0, and are undyed. This suture comes with a CP-2 26mm 1/2c Cutting Edge Reverse Needle and has 36 sutures in a box. Primary code substitute for discontinued code J869H
Catalog No. 50-283-0030 Supplier Med Vet International Supplier No. VCP869H
The Ethicon VCP869H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 2-0, and are undyed. This suture comes with a CP-2 26mm 1/2c Cutting Edge Reverse Needle and has 36 sutures in a box. Primary code substitute for discontinued code J869H