Learn More
Med Vet International Ethicon Coated Vicryl Plus Antibacterial Suture, Size 2-0, CP-1, 27in, 36/Box
Catalog No. 502522154
Encompass_Preferred
The Ethicon VCP466H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 2-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CP-1 reverse cutting needle and has 36 sutures in a box. Primary code substitute for discontinued code J466H
Catalog No. 50-252-2154 Supplier Med Vet International Supplier No. VCP466H
The Ethicon VCP466H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 2-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CP-1 reverse cutting needle and has 36 sutures in a box. Primary code substitute for discontinued code J466H