Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 2-0, ctb-1, 18in, Undyed, 12/Box
Catalog No. 502830025
Encompass_Preferred
The Ethicon VCPB839D suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 18in in length, has a size of 2-0, and are undyed. This suture comes with a CTB-1 36mm 1/2c Blunt Point Ethiguard Needle and has 12 sutures in a box. Primary code substitute for discontinued code JB839
Catalog No. 50-283-0025 Supplier Med Vet International Supplier No. VCPB839D
The Ethicon VCPB839D suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 18in in length, has a size of 2-0, and are undyed. This suture comes with a CTB-1 36mm 1/2c Blunt Point Ethiguard Needle and has 12 sutures in a box. Primary code substitute for discontinued code JB839