Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 2, Tp-1, 54in, Undyed, 24/Box
Catalog No. 502830031
Encompass_Preferred
The Ethicon VCP880T suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 54in in length, has a size of 2, and are undyed. This suture comes with a TP-1 65mm 1/2c Taper Point Needle and has 24 sutures in a box. Primary code substitute for discontinued code J880T
Catalog No. 50-283-0031 Supplier Med Vet International Supplier No. VCP880T
The Ethicon VCP880T suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 54in in length, has a size of 2, and are undyed. This suture comes with a TP-1 65mm 1/2c Taper Point Needle and has 24 sutures in a box. Primary code substitute for discontinued code J880T