Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 3-0, Fs-2, 18in, Violet, 36/Box
Catalog No. 502830511
Encompass_Preferred
The Ethicon VCP393H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 18in in length, has a size of 3-0, and are violet. This suture comes with a FS-2 19mm 3/8c Cutting Edge Reverse Needle and has 36 sutures in a box. Primary code substitute for discontinued code J393H
Catalog No. 50-283-0511 Supplier Med Vet International Supplier No. VCP393H
The Ethicon VCP393H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 18in in length, has a size of 3-0, and are violet. This suture comes with a FS-2 19mm 3/8c Cutting Edge Reverse Needle and has 36 sutures in a box. Primary code substitute for discontinued code J393H