Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 3-0, Sh, 27in, Undyed, 36/Box
Catalog No. 502830717
Encompass_Preferred
The Ethicon VCP416H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 3-0, and are undyed. This suture comes with a SH 26mm 1/2c Taper Needle and has 36 sutures in a box. Primary code substitute for discontinued code J416H
Catalog No. 50-283-0717 Supplier Med Vet International Supplier No. VCP416H
The Ethicon VCP416H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 3-0, and are undyed. This suture comes with a SH 26mm 1/2c Taper Needle and has 36 sutures in a box. Primary code substitute for discontinued code J416H