Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 3-0, Fs-2, 27in, Undyed, 36/Box
Catalog No. 502830692
Encompass_Preferred
The Ethicon VCP423H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 3-0, and are undyed. This suture comes with a FS-2 19mm 3/8c Cutting Edge Reverse Needle and has 36 sutures in a box. Primary code substitute for discontinued code J423H
Catalog No. 50-283-0692 Supplier Med Vet International Supplier No. VCP423H
The Ethicon VCP423H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 3-0, and are undyed. This suture comes with a FS-2 19mm 3/8c Cutting Edge Reverse Needle and has 36 sutures in a box. Primary code substitute for discontinued code J423H