Learn More
Med Vet International Ethicon Vicryl Suture, Size 3-0, CT-1, 27in, 36/Box
Catalog No. 502522363
Encompass_Preferred
The Ethicon J338H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.
Catalog No. 50-252-2363 Supplier Med Vet International Supplier No. J338H
 Restricted Item
Add to Cart
Add to Cart
The Ethicon J338H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.