Learn More
Med Vet International Ethicon Vicryl, 3-0 Polyglactin 910 Suture, 27in SH, 36/Box
Catalog No. 501180836
Encompass_Preferred
The Ethicon J316H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a SH-3 taper point needle and has 36 sutures in a box.
Catalog No. 50-118-0836 Supplier Med Vet International Supplier No. J316H
The Ethicon J316H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 3-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a SH-3 taper point needle and has 36 sutures in a box.