Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 5-0, P-2, 18in, Undyed, 12/Box
Catalog No. 502830691
Encompass_Preferred
The Ethicon VCP503G suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 18in in length, has a size of 5-0, and are undyed. This suture comes with a P-2 8mm 1/2c Cutting Edge Prime Reverse Needle and has 12 sutures in a box. Primary code substitute for discontinued code J503G
Catalog No. 50-283-0691 Supplier Med Vet International Supplier No. VCP503G
The Ethicon VCP503G suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 18in in length, has a size of 5-0, and are undyed. This suture comes with a P-2 8mm 1/2c Cutting Edge Prime Reverse Needle and has 12 sutures in a box. Primary code substitute for discontinued code J503G