Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Plus Antibacterial Suture, Size 5-0, Rb-1, 27in, Undyed, 36/Box
Catalog No. 502830754
Encompass_Preferred
The Ethicon VCP213H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 5-0, and are undyed. This suture comes with a RB-1 17mm 1/2c Taper Needle and has 36 sutures in a box. Primary code substitute for discontinued code J303H
Catalog No. 50-283-0754 Supplier Med Vet International Supplier No. VCP213H
The Ethicon VCP213H suture is a high-quality absorbable Vicryl Polyglactin 910 Plus Antibacterial suture. It measures 27in in length, has a size of 5-0, and are undyed. This suture comes with a RB-1 17mm 1/2c Taper Needle and has 36 sutures in a box. Primary code substitute for discontinued code J303H