Learn More
Med Vet International Ethicon Vicryl Suture, Size 5-0, P-3, 18in, Undyed, 12/Box
Catalog No. 501180848
Encompass_Preferred
The Ethicon J493G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 5-0, they are undyed and typically have an absorption rate of 56-70 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-118-0848 Supplier Med Vet International Supplier No. J493G
 Restricted Item
Add to Cart
Add to Cart
The Ethicon J493G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 5-0, they are undyed and typically have an absorption rate of 56-70 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.