Learn More
Med Vet International Ethicon Vicryl Suture, Size 7-0, P-1, 18in, 12/Box
Catalog No. 502522353
Encompass_Preferred
The Ethicon J488G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 7-0, they are undyed and typically have an absorption rate of 56-70 days. This suture comes with a P-1 Precision Point reverse cutting needle and has 36 sutures in a box.
Catalog No. 50-252-2353 Supplier Med Vet International Supplier No. J488G
The Ethicon J488G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 7-0, they are undyed and typically have an absorption rate of 56-70 days. This suture comes with a P-1 Precision Point reverse cutting needle and has 36 sutures in a box.