Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Suture, Size 1, CT-1, 27in, Undyed, 36/Box
Catalog No. 502522366
Encompass_Preferred
The Ethicon J261H suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 27in in length, has a size of 1, and are undyed. This suture comes with a CT-1 36mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-252-2366 Supplier Med Vet International Supplier No. J261H
The Ethicon J261H suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 27in in length, has a size of 1, and are undyed. This suture comes with a CT-1 36mm 1/2c Taper Needle and has 36 sutures in a box.