Learn More
Med Vet International Ethicon Vicryl Polyglactin 910 Suture, Size 2-0, LIGAPAK REEL, 54in, Violet, 12/Box
Catalog No. 502522367
Encompass_Preferred
The Ethicon J206G suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 54in in length, has a size of 2-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a ligapak Dispensing reel and has 12 sutures in a box.
Catalog No. 50-252-2367 Supplier Med Vet International Supplier No. J206G
The Ethicon J206G suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 54in in length, has a size of 2-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a ligapak Dispensing reel and has 12 sutures in a box.